Filter vials, SINGLE StEP™
PES, 0,2 µm, gris
N° de catalogue 548-1406
Fournisseur: THOMSON INSTRUMENT
Aller à
Product Details & Documents
Simple replacements for HPLC vials, HPLC caps, syringes, needles, and syringe filters. Filter vials minimise steps and loss of product. They protect HPLC, GC, GC-MS and LC-MS systems against contaminants and particulates.
- Compatible with both UHPLC and standard HPLC autosamplers
- Colour coded pre-slit caps for easy identification of membrane and porosity
- Low dead volume unit contains rugged PP insert with 450 µl loading capacity
Spécifications
- Type de membrane:PES
- Taille de pore:0,2 µm
- Couleur:gris
- Seq:04