Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Tubes, PTFE

4,50 mm, 1,00 mm, 6,50 mm
  DENE3404565
Tubes, PTFE
  DENE3404565
Tubes, PTFE
Tubes, PTFE

4,50 mm, 1,00 mm, 6,50 mm

 
SFr. 192.00
   
1 * 5 m
 

PTFE, white, translucent.

  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • Free of extractable substances, physiologically neutral, biocompatible

This tubing is characterised by its non stick surface and excellent dielectric properties.

Temperature resistance -200 to +200 °C.

 
  • Tube Ø ext.:
    6,50 mm
  • Tube Ø int.:
    4,50 mm
  • Épaisseur de paroi:
    1,00 mm
  • Seq:
    19