Power Cables for Vacuum Controllers
Power cable LKG, EU-plug
Catalog # VACU20637652
Supplier: VACUUBRAND
Jump to:
Product Details & Documents
Power cables for Peltronic® exhaust vapour condenser and vacuum controllers VNC 2.
Specifications
- Désignation:Power cable LKG, EU-plug
- Seq:03