Base materials, Millipore®
Base materials, Millipore®
Proteose peptone from animal tissue, for antibiotic production, growth of fastidious microorganisms and production of bacterial toxins, 1 kg
N° de catalogue SIALP0431-1KG
Fournisseur:   Merck
Base materials, Millipore®
N° de catalogue SIALP0431-1KG
Fournisseur:  Merck
Numéro CAS:  
Base materials, Millipore®
Proteose peptone from animal tissue, for antibiotic production, growth of fastidious microorganisms and production of bacterial toxins, 1 kg
Numéro CAS:  
Prix
402.00 SFr.
Connexion Pour la disponibilité
1 * 1 kg
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Documents
Spécifications
Spécifications
  • Cdt:
    1 kg
  • Désignation:
    Proteose peptone from animal tissue, for antibiotic production, growth of fastidious microorganisms and production of bacterial toxins