Accessoires
Accessoires
Glass stage insert (D = 110 mm; d = 38 mm)
Catalog # ZEIS4155101302000
Supplier:   ZEISS
Accessoires
Catalog # ZEIS4155101302000
Supplier:  ZEISS
Accessoires
Accessoires
Glass stage insert (D = 110 mm; d = 38 mm)
Price
SFr. 86.45
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Désignation:
    Glass stage insert (D = 110 mm; d = 38 mm)
  • Seq:
    05