Detergent removal system, DetergentOUT™ Tween®
DetergentOUT™ Tween®, Medi, 10
Fournisseur: G-Biosciences
Detergent removal system, DetergentOUT™ Tween®
N° de catalogue 786-215
Fournisseur: G-Biosciences
Numéro CAS:
Aller à
Product Details & Documents
DetergentOUT™ Tween® is a spin column format detergent removal resin for polysorbate or Tween® detergents or surfactants.
DetergentOUT™ Tween® specifically removes polysorbate detergents without significant loss of proteins, dilution of the protein solution, or change to the buffer composition of the protein solution.
Documents
Spécifications
Spécifications
- Cdt:10
- Désignation:DetergentOUT™ Tween®, Medi