Retort rings
Ring with bosshead, 236 g, 100 mm
N° de catalogue JUCHVL04960
Fournisseur: JUCHHEIM Laborgerate GmbH
Aller à
Product Details & Documents
Steel, epoxy coated.
- Available with or without bosshead
- Bosshead is made of malleable iron
Spécifications
- Ø ext.:100 mm
- Désignation:Ring with bosshead, 236 g
- Seq:09