Retort rings
Ring without bosshead, 157 g, 70 mm
N° de catalogue JUCHVL04900
Fournisseur: JUCHHEIM Laborgerate GmbH
Aller à
Product Details & Documents
Steel, epoxy coated.
- Available with or without bosshead
- Bosshead is made of malleable iron
Spécifications
- Ø ext.:70 mm
- Désignation:Ring without bosshead, 157 g
- Seq:02