Polystyrene standards, GPC/SEC columns calibration
Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 01 g, 1
N° de catalogue VARIPL2013-5001
Fournisseur: VARIAN
Polystyrene standards, GPC/SEC columns calibration
N° de catalogue VARIPL2013-5001
Fournisseur: VARIAN
Numéro CAS:
Aller à
Product Details & Documents
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
- Compatible with most organic solvents
- Certificate of analysis meets international protocols
- Calibration capability for virtually all applications
Documents
Certificats
Rechercher par numéro de lotSpécifications
Spécifications
- Cdt:1
- Désignation:Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,02, quantities 01 g