Sample vials, closed-top
40,00 ml, incolore, Oui, Closed, 24-414, PTFE-faced
N° de catalogue EPSCC336-0040
Fournisseur: EP SCIENTIFIC
Sample vials, closed-top
N° de catalogue EPSCC336-0040
Fournisseur: EP SCIENTIFIC
Aller à
Product Details & Documents
Clear or amber sample vials made from borosilicate glass.
- Designed for water sample collection for volatile organic analysis (VOA)
- Optimised for use with automation
- With closed-top white PP cap bonded with PTFE-lined silicone septum for secure sealing
Processed or certified to meet or exceed EPA Performance Based Specifications for volatile organic analysis with certificate. Processed vials are processed in the same manner as certified vials and come with all the features of certified vials, but vials are not barcoded and CoA is not included.
Spécifications
- Avec bouchon:Oui
- Taille du bouchon:24-414
- Septum:PTFE-faced
- Capacité:40,00 ml
- Couleur:incolore
- Seq:06
- Type de capuchon:Closed
- Cat. No.:EPSCC336-0040
- Certification environnementale:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..