Ferrules and nuts for Thermo Scientific instruments
Ferrules and nuts for Thermo Scientific instruments
For use with TRACE Cold on-column injectors, 0,1 – 0,25 mm
N° de catalogue 555-9051
Fournisseur:   Thermo Fisher Scientific
Ferrules and nuts for Thermo Scientific instruments
N° de catalogue 555-9051
Ferrules and nuts for Thermo Scientific instruments
Ferrules and nuts for Thermo Scientific instruments
For use with TRACE Cold on-column injectors, 0,1 – 0,25 mm
Prix
SFr. 206.00
Connexion Pour la disponibilité
1 * 10 ST
Product Details & Documents
Spécifications
  • Ø trou:
    0,1 – 0,25 mm
  • Désignation:
    For use with TRACE Cold on-column injectors
  • Seq:
    04