Extender tools
Extender tool, Aluminium, Standard knurl, short
N° de catalogue UPCHP-399
Fournisseur: UPCHURCH SCIENTIFIC
Aller à
Product Details & Documents
These tools made from aluminium or Delrin® can be used to tighten most knurled nuts in hard to reach places. For precise tightening, the extender tools listed with ¼" hex drives are designed to adapt to any torque wrench with a female ¼" socket.
Spécifications
- Désignation:Extender tool
- Matière:Aluminium
- Seq:04
- À utiliser avec une tête de type:Standard knurl, short