Jars with overlapping lid
30 mm, 16 mm
Catalog # SEMA4685
Supplier: SEMADENI
Jump to:
Product Details & Documents
Jars, made of PS, clear, with closing lid.
- Good chemical resistance
- Easy to clean
Suitable for ointments, powders, and granules.
Specifications
- Ø ext.:30 mm
- Hauteur:16 mm
- Seq:06



