Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Jars with overlapping lid

Jars with overlapping lid

19 mm, 12 mm
N° de catalogue SEMA4684
Fournisseur:   SEMADENI
Jars with overlapping lid
N° de catalogue SEMA4684
Fournisseur:  SEMADENI
Jars with overlapping lid
Jars with overlapping lid

19 mm, 12 mm

Prix
55.50 SFr.
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents

Jars, made of PS, clear, with closing lid.

  • Good chemical resistance
  • Easy to clean

Suitable for ointments, powders, and granules.

Spécifications
  • Ø ext.:
    19 mm
  • Hauteur:
    12 mm
  • Seq:
    04