Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Jars with overlapping lid

Jars with overlapping lid

46 mm, 42 mm
N° de catalogue SEMA2055
Fournisseur:   SEMADENI
Jars with overlapping lid
N° de catalogue SEMA2055
Fournisseur:  SEMADENI
Jars with overlapping lid
Jars with overlapping lid

46 mm, 42 mm

Prix
76.20 SFr.
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents

Jars, made of PS, clear, with closing lid.

  • Good chemical resistance
  • Easy to clean

Suitable for ointments, powders, and granules.

Spécifications
  • Ø ext.:
    46 mm
  • Hauteur:
    42 mm
  • Seq:
    12
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.