Draining racks, wall mounted
Draining racks, wall mounted
Draining rack, with 16 rods of 200 mm and 16 rods of 140 mm, 450×600 mm
N° de catalogue SEMA2110
Fournisseur:   SEMADENI
Draining racks, wall mounted
N° de catalogue SEMA2110
Fournisseur:  SEMADENI
Draining racks, wall mounted
Draining rack, with 16 rods of 200 mm and 16 rods of 140 mm, 450×600 mm
Prix
226.00 SFr.
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
White, PVC drain channel, polyethylene coated metal rack.
  • Drain channels inclined toward the centre and provided with a drain point
  • Metal frame and drip plasticised PE rods
  • The rack is hung on the wall plate and can be easily removed for cleaning
Spécifications
  • Désignation:
    Draining rack, with 16 rods of 200 mm and 16 rods of 140 mm
  • Seq:
    02
  • l×h:
    450×600 mm