Pipette tips, multi crystal
0,5 - 10 µl, 10 racks, 96 each
Catalog # SORE22220
Supplier: Sorenson Bioscience
Jump to:
Product Details & Documents
Virgin PP, non sterile, transparent.
- Conical top and smooth narrow lumen for maximum sample recovery and accuracy
- Manufactured from virgin materials and guaranteed free from pyrogens and heavy metals
- Certified free from RNases, DNases, human DNA and PCR inhibitors
These 10 μl pipette tips are optimised to fit Eppendorf pipettes.
Specifications
- Emballage:10 racks, 96 each
- Volume:0,5 - 10 µl
- Seq:02