Pipette tips, multi crystal
Pipette tips, multi crystal
0,5 - 10 µl, Bulk (1000/bag)
N° de catalogue SORE17000
Fournisseur:   Sorenson Bioscience
Pipette tips, multi crystal
N° de catalogue SORE17000
Fournisseur:  Sorenson Bioscience
Pipette tips, multi crystal
Pipette tips, multi crystal
0,5 - 10 µl, Bulk (1000/bag)
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1.000 ST
Product Details & Documents
Virgin PP, non sterile, transparent.
  • Conical top and smooth narrow lumen for maximum sample recovery and accuracy
  • Manufactured from virgin materials and guaranteed free from pyrogens and heavy metals
  • Certified free from RNases, DNases, human DNA and PCR inhibitors
These 10 μl pipette tips are optimised to fit Eppendorf pipettes.
Spécifications
  • Emballage:
    Bulk (1000/bag)
  • Volume:
    0,5 - 10 µl
  • Seq:
    01