Micro Curettes
Micro curette, tip diameter 2,0 mm, serrated, 125 mm
N° de catalogue FSCI10083-20
Marketsource
Fournisseur: FINE SCIENCE TOOLS
Aller à
Product Details & Documents
Stainless steel. A curette tip consists of a scoop with sharp edges and is used for scraping or cleaning. It also can be used for separating and transporting, inoculating plates or transferring small amounts of chemicals.
Spécifications
- Désignation:Micro curette, tip diameter 2,0 mm, serrated
- Longueur:125 mm