Mayo scissors
Blunt/blunt, curved tips, Toughcut®, 170 mm, 45 mm, +
N° de catalogue FSCI14111-17
Marketsource
Fournisseur: FINE SCIENCE TOOLS
Aller à
Product Details & Documents
These scissors are made of stainless steel.
Spécifications
- Type:Blunt/blunt, curved tips, Toughcut®
- Longueur:170 mm
- Seq:03
- Serrated:+
- Longueur de lame:45 mm