Dumont #5 forceps
#5, Dumoxel, 110 mm, Biology, Straight, 0,05×0,02 mm
Catalog # FSCI11252-30
Marketsource
Supplier: FINE SCIENCE TOOLS
Jump to:
Product Details & Documents
Specifications
- Type:#5
- Longueur:110 mm
- Matière:Dumoxel
- Seq:05
- Style de pointe:Biology
- Tip size:0,05×0,02 mm
- Tip shape:Straight