Frocks, PP
blanc, XL
N° de catalogue HOPE40.301/04
Fournisseur: Medicom
Aller à
Product Details & Documents
These single-use hygiene frocks made from PP protect the working environment from particles coming from the operator's body (dander, hair, perspiration, etc.).
- With collar and long sleeves
- Elasticated cuffs
- Snap buttons
- Without pockets
Weight: 40 g/m²
Documents
Spécifications
- Taille de confection:XL
- Couleur:blanc
- Seq:03
- Cat. No.:HOPE40.301/04
- Poids:40 g/m²
- Type de fermeture:Avec boutons pression
- Applications des tenues:À usage universel|Industriel
- Poches:sans poches