Stoppers, PTFE
Stoppers, PTFE
blanc, 34/35
N° de catalogue BOHLH936-08
Fournisseur:   Bohlender
Stoppers, PTFE
N° de catalogue BOHLH936-08
Fournisseur:  Bohlender
Stoppers, PTFE
Stoppers, PTFE
blanc, 34/35
Prix
62.60 SFr.
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
For closing ground joint parts. With ground joint and sealing rings on the outside; knurled or hexagonal grip. Compared with glass stoppers, they are easily removable and can be used without grease.
  • PTFE
  • Temperature resistance from −200 to + 250 °C
  • Chemical resistance
  • FDA conform
The stoppers can expand under heat which might lead to a breaking of the ground joint sockets.
Spécifications
  • RN:
    34/35
  • Couleur:
    blanc