Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Tubes, PTFE

10,00 mm, 1,00 mm, 12,00 mm
  DENE3401012
Tubes, PTFE
  DENE3401012
Tubes, PTFE
Tubes, PTFE

10,00 mm, 1,00 mm, 12,00 mm

 
SFr. 345.00
   
1 * 5 m
 

PTFE, white, translucent.

  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • Free of extractable substances, physiologically neutral, biocompatible

This tubing is characterised by its non stick surface and excellent dielectric properties.

Temperature resistance -200 to +200 °C.

 
  • Tube Ø ext.:
    12,00 mm
  • Tube Ø int.:
    10,00 mm
  • Épaisseur de paroi:
    1,00 mm
  • Seq:
    29
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.