Accessoires
Preconditioning cartridge holder: to ensure optimum performance when feedwater is > 1 µS/cm
For PURELAB® flex 2 only
For PURELAB® flex 2 only
Catalog # 171-0353
Jump to:
Discontinued
Product Details & Documents
Specifications
- Désignation:Preconditioning cartridge holder: to ensure optimum performance when feedwater is > 1 µS/cm
For PURELAB® flex 2 only - Seq:13