Threaded couplings
GL 14
N° de catalogue BOHLH900-01
Fournisseur: Bohlender
Aller à
Product Details & Documents
Connector, screw thread coupling, made of PTFE. Two screw caps made of PPS or PBTP with GL thread and a PTFE/FPM gasket. The product is only exposed to PTFE.
- Applicable for connecting two GL threads of different sizes
- Temperature resisitance from −200 to +250 ºC
- Very good chemical resistance
- FDA conformity
Spécifications
- Filetage:GL 14