VWR® Basic, Frocks, SMS
blanc, M
N° de catalogue 113-1157
Fournisseur: VWR Collection
Aller à
Product Details & Documents
These SMS frocks are manufactured from a latex-free, multi-layer spunbonded - melt-blown - spunbonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
- Snap collar, raglan sleeves, snap closure
- Fluid and particulate barrier
- Light chemical splash resistance
Colour: White or blue
Documents
Spécifications
- Taille de confection:M
- Couleur:blanc
- Seq:02
- Cat. No.:113-1157
- Type de fermeture:Avec boutons pression
- Applications des tenues:Salle propre|Laboratoire
- Genre:Unisex
- Poches:sans poches