VWR® Basic, Frocks, SMS
blanc, XL
  113-1159
VWR® Basic, Frocks, SMS
  113-1159
VWR® Basic, Frocks, SMS
VWR® Basic, Frocks, SMS
blanc, XL
SFr. 209.00
1 * 30 ST
These SMS frocks are manufactured from a latex-free, multi-layer spunbonded - melt-blown - spunbonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
  • Snap collar, raglan sleeves, snap closure
  • Fluid and particulate barrier
  • Light chemical splash resistance
Colour: White or blue
  • Taille de confection:
    XL
  • Couleur:
    blanc
  • Seq:
    04
  • Cat. No.:
    113-1159
  • Type de fermeture:
    Avec boutons pression
  • Applications des tenues:
    Salle propre|Laboratoire
  • Genre:
    Unisex
  • Poches:
    sans poches