Fork wrenches, BOLA
Fork wrench for thread GL 32/GL 45, Filbre de verre/Polyamide
N° de catalogue BOHLD647-24
Fournisseur: Bohlender
Aller à
Product Details & Documents
These glass fibre reinforced PA fork wrenches can be used for tightening or opening screw joints also at high working temperatures.
- Good chemical resistance
- Temperature resistant from −10 to +140 °C
- Low weight reduces risk of injury or damage
Spécifications
- Désignation:Fork wrench for thread GL 32/GL 45
- Matière:Filbre de verre/Polyamide
- Seq:02