ContactUsLinkComponent
Snap Cap, 11mm, Red Silicone/PTFE Single Slit Septa, 100 Stk OMNILAB 0 1 * 100 ST
Catalog # BROW153336
Snap Cap, 11mm, Red Silicone/PTFE Single Slit Septa, 100 Stk OMNILAB 0 1 * 100 ST
Catalog # BROW153336
Frequently Bought Together