Micro capillaries, disposable, Kimble®
Capillary pipettes, 5 µl, blanc
  SCERSP71900-5
Micro capillaries, disposable, Kimble®
  SCERSP71900-5
Micro capillaries, disposable, Kimble®
Micro capillaries, disposable, Kimble®
Capillary pipettes, 5 µl, blanc
636.00 SFr.
1 * 1.000 ST
These disposable micro capillary pipettes are made from soda lime glass, USP Type III.
  • Calibrated to contain
  • Colour-coded
  • Marked with single black capacity ring
  • Packaged in easy-open cylinders for convenient dispensing
 : Designed from ASTM Specification E672.
 : Each cylinder contains 250 pipettes and one pipette device.
  • Code couleur:
    blanc
  • Capacité:
    5 µl
  • Désignation:
    Capillary pipettes
  • Seq:
    01