Drum taps, brass
Self-closing with flame guard, PTFE seal and trailer fitting, Non-corrosive liquids <30 SAE, 3/4"
N° de catalogue MSGA2799
Fournisseur: asecos
Aller à
Product Details & Documents
Brass taps for drums with screw thread.
- Self closing
- Flame guard
- PTFE seal
Informations de commande: MSGA2799 includes a trailer fitting for decanting vessels.
Spécifications
- Filetage:3/4"
- Désignation:Self-closing with flame guard, PTFE seal and trailer fitting
- Seq:01
- Pour:Non-corrosive liquids <30 SAE