Power Cables for Vacuum Controllers
Power cable, Y-shaped KG and LKG, EU-plug
N° de catalogue VACU20636273
Fournisseur: VACUUBRAND
Aller à
Product Details & Documents
Power cables for Peltronic® exhaust vapour condenser and vacuum controllers VNC 2.
Spécifications
- Désignation:Power cable, Y-shaped KG and LKG, EU-plug
- Seq:02