Water purification systems, Select Analyst
Product Details & Documents
The Select Analyst is a compact unit ideal for Laboratory use, delivering a steady supply of > 1MΩ.cm pure water for Laboratory tasks using carbon pre-treatment, reverse osmosis and deionisation.
- RO removes > 98% minerals and > 99% bacteria
- RO boost pump fitted as standard
- LCD colour touch screen panel, visual and audible alarms
- Available with or without integral 20 litre storage
- Semi-automatic clean cycle
- USB port to download event data and upload software updates
- WiFi connection
Typical applications include glassware rinsing, buffers and stains, reagent make-up, and media preparation.
Installation kit and all consumables included for first year's operation.
Delivery information: Supplied with alphanumeric display panel, pretreatment cartridge, reverse osmosis module, deionisation module and single tank bib-tap. External tanks required for GP models are also available, please contact your local Avantor office for details.
Specifications
- Désignation:Select Analyst 40 IT with boost pump and internal tank (20 L)
- Seq:02
- Cat. No.:171-0183
- Débit:3,6 - 6 L/h
- Teneur COT:<50 ppb
- Poids:28 - 43 kg
- Eau d'alimentation:Potable (tap)
- Bactéries:>99% rejection*
- L×P×H:440×560×750 mm
- Inorganiques @ 25°C:>1 MΩ/cm
- Modèle:Analyst 40
Accessories
Product Information
- Catalog #DésignationAvailabilityPrice
- 171-0614External tank wall mounting kit, for Select Analyst and HP systemsLog in for availability1 * 1 ST - SFr. 1’790.00
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
- 171-0256Annual consumable pack for Select Analyst 320/Purewater 300 containing 2 x PT16 and 2 x PP16 cartridgesLog in for availability1 * 1 ST - SFr. 5’260.00
Product Information
- Catalog #DésignationAvailabilityPrice
- 171-0265Filter element, 0,2 µm for current Select range (with grey door) and previous L2G Select range (with blue door), used in HP, Fusion, Neptune water purification systemsLog in for availability1 * 1 ST - SFr. 981.00
Product Information
- Catalog #DésignationAvailabilityPrice
- 171-0264Replacement dispense gun for water purification systemsLog in for availability1 * 1 ST - SFr. 934.00
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



