Inlet septa
Septa, general purpose, grey, 9,5 mm
Katalognr. HEWL5080-8728-50
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
- Made up low bleed silicone rubber formulations
- Efficient by using sharp syringe needles
- Septa should be replaced to avoid leaks and sample loss
- To be used with recommended temperature range
Technische Daten
- Ø:9,5 mm
- Désignation:Septa, general purpose, grey