Pointes à filtre, aeroject®
Extended, 0,1 - 10 µl, +, Pipettes couramment utilisées, 10 portoirs, 96 cônes chacun
N° de catalogue RATI1770010
Fournisseur: RATIOLAB
Aller à
Product Details & Documents
These sterile pipette tips with filters effectively prevent the pipettor shaft from contamination with aerosols and liquids.
- Free of RNase-, DNase-, DNA-, pyrogens, ATP
- Free of human DNA and PCR inhibitors
- Tips and aerosol filters made of ultra-hydrophobic PE-HD
- Minimised retention due to special surface treatment
- Perfect and tight fit on all major pipettor brands
- Tips with extended length: Ideal for operation with narrow vessels and reduced risk of cross-contamination
Emballage: Packed in plastic racks with hinged lids.
Spécifications
- Emballage:10 portoirs, 96 cônes chacun
- Stérile:+
- Volume:0,1 - 10 µl
- Style de pointe:Extended
- Pour l'utilisation avec:Pipettes couramment utilisées