HyClone™ CDM4HEK293
CDM4HEK293 medium without phenol red, L-glutamine, HEPES, Liquide, 1 L
Fournisseur: HyClone products (Cytiva)
Aller à
Product Details & Documents
HyClone™ CDM4HEK293 media are chemically-defined, animal-derived component free (ADCF) cell culture media.
- Allows for direct or sequential adaptation
- Component traceability
- Manufactured under cGMP
- Metabolically designed for high cell yield and recombinant protein/vector production
HyClone CDM4HEK293 is a regulatory-friendly medium developed through the Metabolic Pathway Design to support the extremely high productivity and cell density in suspension cultures.
Documents
Spécifications
Spécifications
- Format du milieu:Liquide
- Cdt:1 L
- Désignation:CDM4HEK293 medium without phenol red, L-glutamine, HEPES