ContactUsLinkComponent
Bote 10 x 10, noir, 140 x 153 x 53 mm, compartiments de 12,5 x 12,5 mm, en polypropylne, autoclavable 121c et congelable -90 c 1 * 1 Pce
Katalognr. 479-1527
Supplier:  GLW
undefined
Bote 10 x 10, noir, 140 x 153 x 53 mm, compartiments de 12,5 x 12,5 mm, en polypropylne, autoclavable 121c et congelable -90 c 1 * 1 Pce
Katalognr. 479-1527
Supplier:  GLW
Frequently Bought Together