Tubes and systems for active sampling
Sampling tubes amines, TEA
N° de catalogue DRAE8101271
Fournisseur: Dräger
Aller à
Product Details & Documents
Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analyses.
- High precision guaranteed
- Very reliable in use
- Improved colour depth and contrast using an optimised production method
Spécifications
- Type:TEA
- Désignation:Sampling tubes amines
- Seq:17