Vial, Klarglas, 15 x 45 mm 100 Stk./Pkg. OMNILAB 0 1 * 100 ST
BROW150745
Vial, Klarglas, 15 x 45 mm 100 Stk./Pkg. OMNILAB 0 1 * 100 ST
BROW150745
Frequently Bought Together
VWR®, Test Tubes without Rim
: VWR Collection
Acétone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Mouchoirs en papier
: VWR Collection
VWR®, Gants en nitrile
: VWR Collection
Lead-free steam indicator tapes, Comply™
: 3M MEDICAL
VWR®, Pipettes de transfert, High Performance
: VWR Collection
[EN]SCREW CAPS 13-425 W. CORK/PTFE LINER 1 * 100 ST
: BROWN CHROMATOGRAPHY SUPPLIES
Anti-CHST13 Rabbit Polyclonal Antibody
: ProSci Inc.
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF