Rack overlays for sample racks
Rack overlay kit for 16 mm O.D. tube rack (allows 60, 13 mm OD tubes to be used in the rack)
N° de catalogue VARI6610026100
Fournisseur: VARIAN
Aller à
Product Details & Documents
Spécifications
- Désignation:Rack overlay kit for 16 mm O.D. tube rack (allows 60, 13 mm OD tubes to be used in the rack)
- Seq:01