Normal cell line blots, human
Normal skin cell line blot, human, 1
N° de catalogue TB52
Fournisseur: G-Biosciences
Aller à
Product Details & Documents
Ready to screen, normal tissue human cell line blots from epithelial, endothelial, skin, and muscle tissue.
These premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using western transfer buffer. These blots are ready to be blocked and probed with antibodies of choice.
Spécifications
Spécifications
- Cdt:1
- Désignation:Normal skin cell line blot, human