Normal cell line blots, human
Normal cell line blots, human
Normal epithelial cell line blot, human, 1
N° de catalogue TB50
Fournisseur:   G-Biosciences
Normal cell line blots, human
N° de catalogue TB50
Fournisseur:  G-Biosciences
Numéro CAS:  
Normal cell line blots, human
Normal cell line blots, human
Normal epithelial cell line blot, human, 1
Numéro CAS:  
Prix
SFr. 1’170.00
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Ready to screen, normal tissue human cell line blots from epithelial, endothelial, skin, and muscle tissue.
These premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using western transfer buffer. These blots are ready to be blocked and probed with antibodies of choice.
Spécifications
Spécifications
  • Cdt:
    1
  • Désignation:
    Normal epithelial cell line blot, human