Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Jars with overlapping lid | Avantor

Jars with overlapping lid

27 mm, 10 mm
Katalognr. SEMA1994
Anbieter:   SEMADENI
Jars with overlapping lid
Katalognr. SEMA1994
Anbieter:  SEMADENI
Jars with overlapping lid
Jars with overlapping lid

27 mm, 10 mm

Preis
33.50 SFr.
Einloggen für Verfügbarkeit
1 * 100 ST
Product Details & Documents

Jars, made of PS, clear, with closing lid.

  • Good chemical resistance
  • Easy to clean

Suitable for ointments, powders, and granules.

Technische Daten
  • Ø ext.:
    27 mm
  • Hauteur:
    10 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.