Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Laboratory bowls

1 l, 90 mm, 175 mm
  BOCH8601
 :   BOCHEM
Laboratory bowls
  BOCH8601
 :  BOCHEM
Laboratory bowls
Laboratory bowls

1 l, 90 mm, 175 mm

 
SFr. 24.00
   
1 * 1 ST
 

Bowls are high form, and made of stainless steel 18/10.

  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean

Round multipurpose dishes for a wide range of uses.

 
  • Ø ext.:
    175 mm
  • Capacité:
    1 l
  • Hauteur:
    90 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.