Labelling tapes
Labelling tapes, blanc, 19,0 mm
N° de catalogue SEMA0352
Fournisseur: SEMADENI
Aller à
Product Details & Documents
Labeling tape for glass, plastic, porcelain or metal.
- Tear resistant
- Can be easily written on
- Adhesive strength at low temperature
Length: 66,0 m
Different colours available.
: Do not autoclave.
Spécifications
- Largeur:19,0 mm
- Couleur:blanc
- Désignation:Labelling tapes
- Seq:01
- Cat. No.:SEMA0352