Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Saisie de commande
Login
/
Register
Produits
Services
Solutions numériques
Applications
Support
À propos
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
Lien pour commandes
ContactUsLinkComponent
Home
Tests microbiologiques
Tests sur les anticorps
Ag-AK, latex
Tests latex
Tests latex
Print
Tests latex
Staphaurex Test kit, 120 Tests
Catalog # OXOIR30859901
Certificates
Supplier:
THERMO FISHER DIAGNOSTICS
Tests latex
Catalog # OXOIR30859901
Supplier:
THERMO FISHER DIAGNOSTICS
CAS Number:
Certificates
Expand image
Return to Top
Tests latex
Staphaurex Test kit, 120 Tests
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
SFr. 190.00
Log in
for availability
Unit of Measure
1 * 120 Tests
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
Specifications
Specifications
Cdt:
120 Tests
Désignation:
Staphaurex Test kit
Specifications
Accessories for buchi melting point apparatus M560 and M565
#: 621-0046
Supplier: BUCHI