Hoffmann clips
40 mm
N° de catalogue MURRL141/05
Fournisseur: MURRAY S
Aller à
Product Details & Documents
Bunsen screw clip, nickel plated brass fitted with protection plate and hinged bottom bar.
- Spanning width = Width of gate
Spécifications
- Largeur de serrage:40 mm
- Seq:05
- Cat. No.:MURRL141/05