Hoffmann clips
13 mm
N° de catalogue MURRL141/01
Fournisseur: MURRAY S
Aller à
Product Details & Documents
Bunsen screw clip, nickel plated brass fitted with protection plate and hinged bottom bar.
- Spanning width = Width of gate
Spécifications
- Largeur de serrage:13 mm
- Seq:01