Density bottles, adjusted
5 ml
N° de catalogue LENZ05630210
Fournisseur: Lenz Laborglas
Aller à
Product Details & Documents
Borosilicate glass 3.3. DURAN®, Gay-Lussac type.
- The density bottles (pycnometers) are individually engraved with the actual capacity, with an accuracy of 0,001 cm³
- The top of stopper is ground and polished for ease of wiping clean
- Stopper is size NS 10/19 with capillary
* DIN ISO 3507
Documents
BSE TSE Glass Columns
Certificats BSE/TSE
Spécifications
- Volume nominal:5 ml